Assay Details
Target Gene Details
Entrez Gene ID: | 54606 |
Gene Name: | DEAD-box helicase 56 |
Gene Aliases: |
DDX21, DDX26, NOH61 |
Location: |
Chr.7:44565417-44574538 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX56 | NM_001257189.1 | NP_001244118.1 | ||
| NM_019082.3 | NP_061955.1 | |||
| AF247666.1 | AAG36876.1 | |||
| AJ131712.1 | CAB87992.1 | |||
| AK022774.1 | BAB14238.1 | |||
| AK125595.1 | ||||
| AK222662.1 | BAD96382.1 | |||
| AK315363.1 | ||||
| AL136700.1 | CAB66635.1 | |||
| BC001235.1 | AAH01235.1 | |||
| CR457213.1 | CAG33494.1 |
More Information
Set Membership: |
|

Genomic Map