Assay Details
Target Gene Details
Entrez Gene ID: | 27297 |
Gene Name: | CGRP receptor component |
Gene Aliases: |
CGRP-RCP, CGRPRCP, RCP, RCP9 |
Location: |
Chr.7:66114818-66154568 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRCP | NM_001040647.1 | NP_001035737.1 | ||
| NM_001040648.1 | NP_001035738.1 | |||
| NM_001142414.1 | NP_001135886.1 | |||
| NM_014478.4 | NP_055293.1 | |||
| NR_024548.1 | ||||
| AF073792.1 | AAC25992.1 | |||
| AK290553.1 | ||||
| AK303737.1 | ||||
| AK311829.1 | ||||
| BC040107.1 | AAH40107.1 | |||
| BC105808.1 | AAI05809.1 | |||
| BI600330.1 | ||||
| BX647867.1 | CAI46069.1 | |||
| DA821665.1 | ||||
| DA955978.1 | ||||
| DB472426.1 | ||||
| U51134.1 | AAD05036.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761079 | Chr.7:65601260 - 66224746 on Build GRCh38 | Gain+Loss |
|
| nsv511363 | Chr.7:66122550 - 66132354 on Build GRCh38 | Loss |
|
| esv2664145 | Chr.7:65814760 - 66395282 on Build GRCh38 | Deletion |
|
| dgv11433n54 | Chr.7:66125574 - 66134209 on Build GRCh38 | Loss |
|
| nsv1021186 | Chr.7:66027614 - 66198451 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map