Assay Details
Target Gene Details
Entrez Gene ID: | 79872 |
Gene Name: | Cbl proto-oncogene like 1 |
Gene Aliases: |
HAKAI, RNF188 |
Location: |
Chr.7:107743497-107764997 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBLL1 | NM_001284291.1 | 6 | 2594 | NP_001271220.1 |
| NM_024814.3 | 6 | 2597 | NP_079090.2 | |
| NR_024199.2 | 7 | 2670 | ||
| XM_011516580.2 | 7 | 2867 | XP_011514882.1 | |
| XM_017012643.1 | 6 | 2398 | XP_016868132.1 | |
| XM_017012644.1 | 6 | 2788 | XP_016868133.1 | |
| XM_017012645.1 | 7 | 2722 | XP_016868134.1 | |
| AK293352.1 | ||||
| CN274642.1 | 1 | 305 | ||
| CN291556.1 |
More Information
Set Membership: |
|

Genomic Map