Assay Details
Target Gene Details
Entrez Gene ID: | 10061 |
Gene Name: | ATP binding cassette subfamily F member 2 |
Gene Aliases: |
ABC28, EST133090, HUSSY-18, HUSSY18 |
Location: |
Chr.7:151207837-151227374 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCF2 | NM_005692.4 | NP_005683.2 | ||
| NM_007189.2 | NP_009120.1 | |||
| XM_017011639.1 | XP_016867128.1 | |||
| XM_017011640.1 | XP_016867129.1 | |||
| AF261091.1 | AAG13902.1 | |||
| AF261092.1 | AAG13903.1 | |||
| AJ005016.1 | CAA06290.1 | |||
| AL050291.1 | CAB43392.1 | |||
| BC001661.2 | AAH01661.1 | |||
| BT007451.1 | AAP36119.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv609035 | Chr.7:151199137 - 151258679 on Build GRCh38 | Loss |
|
| esv2735396 | Chr.7:151211690 - 151218511 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map