Assay Details
Target Gene Details
Entrez Gene ID: | 55610 |
Gene Name: | VPS50, EARP/GARPII complex subunit |
Gene Aliases: |
CCDC132, VPS54L |
Location: |
Chr.7:93232340-93361123 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS50 | NM_001257998.1 | NP_001244927.1 | ||
| NM_017667.3 | NP_060137.2 | |||
| XM_011516395.2 | XP_011514697.1 | |||
| XM_011516396.2 | XP_011514698.1 | |||
| XM_017012412.1 | XP_016867901.1 | |||
| AB058764.2 | BAB47490.2 | |||
| AB100163.1 | ||||
| AK000104.1 | BAA90948.1 | |||
| AK027234.1 | BAB15701.1 | |||
| AK055965.1 | ||||
| AK126478.1 | ||||
| AK299582.1 | ||||
| AL832393.1 | ||||
| AL833112.1 | CAI46135.1 | |||
| BC152424.1 |
More Information
Set Membership: |
|

Genomic Map