Assay Details
Target Gene Details
Entrez Gene ID: | 64111 |
Gene Name: | neuropeptide VF precursor |
Gene Aliases: |
C7orf9, RFRP |
Location: |
Chr.7:25224570-25228486 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPVF | NM_022150.3 | NP_071433.3 | ||
| AB040290.1 | BAB17674.1 | |||
| AF330057.1 | AAK94201.1 | |||
| AF440392.1 | AAL90453.1 | |||
| BM714303.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv606424 | Chr.7:24942910 - 25426902 on Build GRCh38 | Gain |
|
| nsv606426 | Chr.7:25214506 - 25581959 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map