Assay Details
Target Gene Details
Entrez Gene ID: | 79872 |
Gene Name: | Cbl proto-oncogene like 1 |
Gene Aliases: |
HAKAI, RNF188 |
Location: |
Chr.7:107743497-107764997 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBLL1 | NM_001284291.1 | NP_001271220.1 | ||
| NM_024814.3 | NP_079090.2 | |||
| NR_024199.2 | ||||
| XM_011516580.2 | XP_011514882.1 | |||
| XM_017012643.1 | XP_016868132.1 | |||
| XM_017012644.1 | XP_016868133.1 | |||
| XM_017012645.1 | XP_016868134.1 | |||
| AK026762.1 | BAB15544.1 | |||
| AK293352.1 | ||||
| AK307992.1 | ||||
| BC027460.2 | AAH27460.2 | |||
| BC130529.1 | ||||
| BC130531.1 | ||||
| BC144176.1 | ||||
| BP201841.1 | ||||
| DB033469.1 |
More Information
Set Membership: |
|

Genomic Map