Assay Details
Target Gene Details
Entrez Gene ID: | 26047 |
Gene Name: | contactin associated protein-like 2 |
Gene Aliases: |
AUTS15, CASPR2, CDFE, NRXN4, PTHSL1 |
Location: |
Chr.7:146116035-148420998 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTNAP2 | NM_014141.5 | NP_054860.1 | ||
| XM_017011950.1 | XP_016867439.1 | |||
| AB020675.1 | ||||
| AF319045.1 | AAK34932.1 | |||
| AK315113.1 | ||||
| BC093780.1 | AAH93780.1 | |||
| BC113373.1 | ||||
| KJ534811.1 |
Target Gene Details
Entrez Gene ID: | 101928700 |
Gene Name: | uncharacterized LOC101928700 |
Gene Aliases: |
- |
Location: |
Chr.7:147080938-147097609 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928700 | NR_110829.1 | 4 | 1847 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034528 | Chr.7:145373994 - 147324855 on Build GRCh38 | Loss |
|
| esv2761380 | Chr.7:147009967 - 147091133 on Build GRCh38 | Gain |
|
| nsv1034510 | Chr.7:147022881 - 147151489 on Build GRCh38 | Loss |
|
| dgv6750n100 | Chr.7:147004988 - 147091121 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map