Assay Details
Target Gene Details
Entrez Gene ID: | 10320 |
Gene Name: | IKAROS family zinc finger 1 |
Gene Aliases: |
CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1 |
Location: |
Chr.7:50303453-50405101 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKZF1 | XM_011515058.2 | XP_011513360.1 | ||
| XM_011515059.2 | XP_011513361.1 | |||
| XM_011515061.2 | XP_011513363.1 | |||
| XM_011515067.2 | XP_011513369.1 | |||
| XM_011515070.2 | XP_011513372.1 | |||
| XM_011515071.2 | XP_011513373.1 | |||
| XM_011515072.2 | XP_011513374.1 | |||
| XM_011515074.2 | XP_011513376.1 | |||
| XM_011515077.2 | XP_011513379.1 | |||
| XM_011515078.2 | XP_011513380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv606897 | Chr.7:49632287 - 50387038 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map