Assay Details
Target Gene Details
Entrez Gene ID: | 55744 |
Gene Name: | cytochrome c oxidase assembly factor 1 homolog |
Gene Aliases: |
C7orf44, MITRAC15 |
Location: |
Chr.7:43608452-43729541 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COA1 | NR_135580.1 | |||
| NR_135581.1 | ||||
| NR_135582.1 | ||||
| NR_135583.1 | ||||
| XM_017012416.1 | XP_016867905.1 | |||
| XM_017012419.1 | XP_016867908.1 | |||
| XM_017012431.1 | XP_016867920.1 | |||
| XM_017012432.1 | XP_016867921.1 | |||
| AK023053.1 | BAB14379.1 | |||
| AK293043.1 | ||||
| BM918741.1 |
Target Gene Details
Entrez Gene ID: | 9263 |
Gene Name: | serine/threonine kinase 17a |
Gene Aliases: |
DRAK1 |
Location: |
Chr.7:43583093-43627379 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK17A | NM_004760.2 | NP_004751.2 | ||
| XM_017012792.1 | XP_016868281.1 | |||
| AB011420.1 | BAA34126.1 | |||
| BC023508.2 | AAH23508.2 | |||
| BC047696.1 | AAH47696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518728 | Chr.7:43555801 - 43614160 on Build GRCh38 | Loss |
|
| esv3612996 | Chr.7:43537785 - 43656356 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142874015] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map