Assay Details
Target Gene Details
Entrez Gene ID: | 105375254 |
Gene Name: | uncharacterized LOC105375254 |
Gene Aliases: |
- |
Location: |
Chr.7:43025325-43114052 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375254 | XM_017012919.1 | XP_016868408.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035016 | Chr.7:43029785 - 43072177 on Build GRCh38 | Loss |
|
| nsv527510 | Chr.7:43038251 - 43080331 on Build GRCh38 | Loss |
|

Genomic Map