Assay Details
Target Gene Details
Entrez Gene ID: | 4885 |
Gene Name: | neuronal pentraxin 2 |
Gene Aliases: |
NARP, NP-II, NP2 |
Location: |
Chr.7:98617285-98629869 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPTX2 | NM_002523.2 | NP_002514.1 | ||
| BC009924.2 | AAH09924.1 | |||
| BC034781.1 | AAH34781.1 | |||
| BC035339.1 | AAH35339.1 | |||
| BC048275.1 | AAH48275.2 | |||
| BI561105.1 | ||||
| U26662.1 | AAA92296.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114610830] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map