Assay Details
Target Gene Details
Entrez Gene ID: | 10142 |
Gene Name: | A-kinase anchoring protein 9 |
Gene Aliases: |
AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO |
Location: |
Chr.7:91940867-92110673 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKAP9 | NM_005751.4 | NP_005742.4 | ||
| NM_147185.2 | NP_671714.1 | |||
| XM_017011642.1 | XP_016867131.1 | |||
| XM_017011643.1 | XP_016867132.1 | |||
| XM_017011644.1 | XP_016867133.1 | |||
| XM_017011645.1 | XP_016867134.1 | |||
| XM_017011646.1 | XP_016867135.1 | |||
| XM_017011647.1 | XP_016867136.1 | |||
| XM_017011648.1 | XP_016867137.1 | |||
| XM_017011649.1 | XP_016867138.1 | |||
| XM_017011650.1 | XP_016867139.1 | |||
| XM_017011651.1 | XP_016867140.1 | |||
| XM_017011652.1 | XP_016867141.1 | |||
| XM_017011653.1 | XP_016867142.1 | |||
| XM_017011654.1 | XP_016867143.1 | |||
| AB019691.1 | BAA78718.1 | |||
| AF026245.1 | AAB86384.1 | |||
| AF247727.4 | ||||
| AJ131693.1 | CAB40713.1 | |||
| BC015533.2 | AAH15533.1 | |||
| CR997756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3571985 | Chr.7:91939149 - 91954762 on Build GRCh38 | Loss |
|
| nsv5838 | Chr.7:91942214 - 91987222 on Build GRCh38 | Deletion |
|
| nsv1021101 | Chr.7:91292598 - 92113836 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map