Assay Details
Target Gene Details
Entrez Gene ID: | 115416 |
Gene Name: | mitochondrial assembly of ribosomal large subunit 1 |
Gene Aliases: |
C7orf30, mtRsfA |
Location: |
Chr.7:23298744-23309573 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MALSU1 | NM_138446.1 | NP_612455.1 | ||
| XM_017011731.1 | XP_016867220.1 | |||
| AK127742.1 | ||||
| AK316559.1 | ||||
| BC012331.1 | AAH12331.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464408 | Chr.7:23293751 - 23343516 on Build GRCh38 | Loss |
|
| esv3612541 | Chr.7:23266901 - 23301590 on Build GRCh38 | Gain |
|
| nsv1016316 | Chr.7:23221900 - 23318018 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73279730] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map