Assay Details
Target Gene Details
Entrez Gene ID: | 23382 |
Gene Name: | adenosylhomocysteinase like 2 |
Gene Aliases: |
ADOHCYASE3, IRBIT2 |
Location: |
Chr.7:129225014-129430211 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHCYL2 | NM_001130720.2 | NP_001124192.1 | ||
| NM_015328.3 | NP_056143.1 | |||
| XM_017011906.1 | XP_016867395.1 | |||
| AB020635.1 | ||||
| BC008349.1 | AAH08349.1 | |||
| BC024325.1 | AAH24325.1 | |||
| BG700017.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116315272] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map