Assay Details
Target Gene Details
Entrez Gene ID: | 84132 |
Gene Name: | ubiquitin specific peptidase 42 |
Gene Aliases: |
- |
Location: |
Chr.7:6078743-6161564 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP42 | XM_006715791.2 | XP_006715854.1 | ||
| XM_011515573.1 | XP_011513875.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034916 | Chr.7:5895149 - 6119339 on Build GRCh38 | Gain |
|
| esv22984 | Chr.7:6083632 - 6105086 on Build GRCh38 | Loss |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
| dgv1183e199 | Chr.7:6081264 - 6105811 on Build GRCh38 | Deletion |
|
| nsv5628 | Chr.7:6065831 - 6107517 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map