Assay Details
Target Gene Details
Entrez Gene ID: | 137872 |
Gene Name: | alcohol dehydrogenase, iron containing 1 |
Gene Aliases: |
ADH8, HMFT2263, HOT |
Location: |
Chr.8:66432483-66468809 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADHFE1 | NM_144650.2 | NP_653251.2 | ||
| AK056992.1 | BAB71335.1 | |||
| AK293530.1 | ||||
| AK294114.1 | ||||
| AK300243.1 | ||||
| AK300881.1 | ||||
| AY033237.1 | AAK44223.1 | |||
| BE241883.1 |
More Information
Set Membership: |
|

Genomic Map