Assay Details
Target Gene Details
Entrez Gene ID: | 51050 |
Gene Name: | peptidase inhibitor 15 |
Gene Aliases: |
CRISP8, P24TI, P25TI |
Location: |
Chr.8:74824537-74855044 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PI15 | NM_001324403.1 | NP_001311332.1 | ||
| NM_015886.4 | NP_056970.1 | |||
| BC074931.2 | AAH74931.1 | |||
| BC074932.2 | AAH74932.1 | |||
| BC126290.1 | ||||
| BC126292.1 | ||||
| CR749657.1 | CAH18451.2 | |||
| D45027.1 | BAA25066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891410 | Chr.8:74600899 - 74942290 on Build GRCh38 | Gain |
|
| dgv7241n100 | Chr.8:74778792 - 74838173 on Build GRCh38 | Loss |
|
| esv28358 | Chr.8:74663344 - 75012983 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114999491] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map