Assay Details
Target Gene Details
Entrez Gene ID: | 10671 |
Gene Name: | dynactin subunit 6 |
Gene Aliases: |
WS-3, WS3, p27 |
Location: |
Chr.8:30156297-30183639 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCTN6 | NM_006571.3 | NP_006562.1 | ||
| AK314128.1 | ||||
| BC013175.1 | AAH13175.1 | |||
| BI562535.1 | ||||
| CR456851.1 | CAG33132.1 | |||
| D84145.1 | BAA19984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6139 | Chr.8:30146815 - 30192125 on Build GRCh38 | Deletion |
|
| nsv831277 | Chr.8:29977428 - 30180297 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map