Assay Details
Target Gene Details
Entrez Gene ID: | 286053 |
Gene Name: | NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase |
Gene Aliases: |
C8orf36, MMS21, NSE2, ZMIZ7 |
Location: |
Chr.8:125091818-125367125 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSMCE2 | NM_173685.2 | NP_775956.1 | ||
| XM_005250875.2 | XP_005250932.1 | |||
| XM_005250876.4 | XP_005250933.1 | |||
| XM_011516974.2 | XP_011515276.1 | |||
| XM_011516975.1 | XP_011515277.1 | |||
| XM_017013331.1 | XP_016868820.1 | |||
| XM_017013333.1 | XP_016868822.1 | |||
| AK057002.1 | BAB71338.1 | |||
| BC032797.1 | AAH32797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv465803 | Chr.8:125055955 - 125120574 on Build GRCh38 | Loss |
|
| nsv1022954 | Chr.8:125105569 - 125168089 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map