Assay Details
Target Gene Details
Entrez Gene ID: | 6934 |
Gene Name: | transcription factor 7 like 2 |
Gene Aliases: |
TCF-4, TCF4 |
Location: |
Chr.10:112950219-113167678 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv518n106 | Chr.10:112949441 - 112951941 on Build GRCh38 | Deletion |
|
| nsv1070891 | Chr.10:112949140 - 112951941 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map