Assay Details
Target Gene Details
Entrez Gene ID: | 6229 |
Gene Name: | ribosomal protein S24 |
Gene Aliases: |
DBA3, S24 |
Location: |
Chr.10:78033760-78056813 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS24 | NM_001026.4 | NP_001017.1 | ||
| NM_001142282.1 | NP_001135754.1 | |||
| NM_001142283.1 | NP_001135755.1 | |||
| NM_001142284.1 | NP_001135756.1 | |||
| NM_001142285.1 | NP_001135757.1 | |||
| NM_033022.3 | NP_148982.1 | |||
| AF147330.1 | ||||
| AK311776.1 | ||||
| AK311910.1 | ||||
| BC000523.2 | AAH00523.1 | |||
| BC071926.1 | AAH71926.1 | |||
| BM745909.1 | ||||
| BX537975.1 | CAD97939.1 | |||
| BX647260.1 | ||||
| CD176901.1 | ||||
| CD677556.1 | ||||
| DB116225.1 | ||||
| M31520.1 | AAA36588.1 |
More Information
Set Membership: |
|

Genomic Map