Assay Details
Target Gene Details
Entrez Gene ID: | 4255 |
Gene Name: | O-6-methylguanine-DNA methyltransferase |
Gene Aliases: |
- |
Location: |
Chr.10:129467184-129770983 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGMT | NM_002412.4 | NP_002403.2 | ||
| XM_005252682.2 | XP_005252739.1 | |||
| AK293593.1 | ||||
| BC000824.1 | AAH00824.1 | |||
| BI520029.1 | ||||
| DC387686.1 | ||||
| DC416206.1 | ||||
| M29971.1 | AAA59596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053385 | Chr.10:129464651 - 129478805 on Build GRCh38 | Loss |
|
| nsv1054536 | Chr.10:128923253 - 129746972 on Build GRCh38 | Gain |
|
| esv3577850 | Chr.10:128673756 - 129928631 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map