Assay Details
Target Gene Details
Entrez Gene ID: | 783 |
Gene Name: | calcium voltage-gated channel auxiliary subunit beta 2 |
Gene Aliases: |
CACNLB2, CAVB2, MYSB |
Location: |
Chr.10:18140560-18545881 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNB2 | NM_001167945.1 | NP_001161417.1 | ||
| NM_201571.3 | NP_963865.2 | |||
| NM_201572.3 | NP_963866.2 | |||
| NM_201593.2 | NP_963887.2 | |||
| NM_201596.2 | NP_963890.2 | |||
| NM_201597.2 | NP_963891.1 | |||
| AF137376.1 | AAD33729.1 | |||
| AF423190.1 | AAL16949.1 | |||
| AF423191.1 | AAL16950.1 | |||
| AK293837.1 | ||||
| AK296471.1 | ||||
| AK307526.1 | ||||
| AK311694.1 | ||||
| AL119895.1 | ||||
| AY393858.1 | AAQ97606.1 | |||
| AY393860.1 | AAQ97608.1 | |||
| AY393861.1 | AAQ97609.1 | |||
| AY675091.1 | AAV49501.1 | |||
| BC136409.1 | ||||
| DC307859.1 | ||||
| DC352526.1 | ||||
| S60415.1 | AAB51370.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054616 | Chr.10:18145025 - 18163040 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map