Assay Details
Target Gene Details
Entrez Gene ID: | 132 |
Gene Name: | adenosine kinase |
Gene Aliases: |
AK |
Location: |
Chr.10:74151185-74709303 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADK | NM_001202450.1 | NP_001189379.1 | ||
| NM_006721.3 | NP_006712.2 | |||
| XM_017015698.1 | XP_016871187.1 | |||
| XM_017015700.1 | XP_016871189.1 | |||
| XM_017015704.1 | XP_016871193.1 | |||
| XM_017015705.1 | XP_016871194.1 | |||
| AK301590.1 | ||||
| CB133126.1 | ||||
| DA356691.1 | ||||
| U33936.1 | AAB01689.1 | |||
| U90338.1 | AAB50234.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75502949] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map