Assay Details
Target Gene Details
Entrez Gene ID: | 10659 |
Gene Name: | CUGBP, Elav-like family member 2 |
Gene Aliases: |
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR |
Location: |
Chr.10:10462515-11336675 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Gene Details
Entrez Gene ID: | 439950 |
Gene Name: | CELF2 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.10:11071541-11105488 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELF2-AS2 | NR_046108.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047445 | Chr.10:10789399 - 11147629 on Build GRCh38 | Gain |
|
| nsv5798 | Chr.10:11060892 - 11094347 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map