Assay Details
Target Gene Details
Entrez Gene ID: | 6934 |
Gene Name: | transcription factor 7 like 2 |
Gene Aliases: |
TCF-4, TCF4 |
Location: |
Chr.10:112950219-113167678 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
More Information
Set Membership: |
|

Genomic Map