Assay Details
Target Gene Details
Entrez Gene ID: | 9585 |
Gene Name: | kinesin family member 20B |
Gene Aliases: |
CT90, KRMP1, MPHOSPH1, MPP-1, MPP1 |
Location: |
Chr.10:89701590-89774943 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF20B | NM_001284259.1 | NP_001271188.1 | ||
| NM_016195.3 | NP_057279.2 | |||
| AB033337.1 | BAB69456.1 | |||
| AK024959.1 | BAB15043.1 | |||
| AL117496.1 | CAB55962.1 | |||
| AL137392.1 | CAB70720.1 | |||
| AY282406.1 | AAP40330.1 | |||
| AY282407.1 | AAP40331.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054572 | Chr.10:89712199 - 89777313 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map