Assay Details
Target Gene Details
Entrez Gene ID: | 105 |
Gene Name: | adenosine deaminase, RNA specific B2 (inactive) |
Gene Aliases: |
ADAR3, RED2 |
Location: |
Chr.10:1177313-1737476 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADARB2 | NM_018702.3 | NP_061172.1 | ||
| AF034837.1 | AAF78094.1 | |||
| AK055107.1 | ||||
| AK057763.1 | ||||
| AK289895.1 | ||||
| AK307451.1 | ||||
| AK308579.1 | ||||
| BC137477.1 | ||||
| BC139725.1 | ||||
| BC140852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion |
|
| nsv527883 | Chr.10:1172090 - 1272353 on Build GRCh38 | Gain |
|
| nsv549768 | Chr.10:1120460 - 1308893 on Build GRCh38 | Gain |
|
| esv2761563 | Chr.10:1184503 - 1299763 on Build GRCh38 | Loss |
|
| nsv509340 | Chr.10:1186931 - 1257850 on Build GRCh38 | Insertion |
|
| nsv1161922 | Chr.10:1104778 - 1240765 on Build GRCh38 | Duplication |
|
| dgv655n100 | Chr.10:1133579 - 1495256 on Build GRCh38 | Gain |
|
| dgv656n100 | Chr.10:1149274 - 1245720 on Build GRCh38 | Gain |
|
| nsv1041498 | Chr.10:1071260 - 1717996 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74400961] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map