Assay Details
Target Gene Details
Entrez Gene ID: | 3422 |
Gene Name: | isopentenyl-diphosphate delta isomerase 1 |
Gene Aliases: |
IPP1, IPPI1 |
Location: |
Chr.10:1039419-1056716 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDI1 | NM_001317955.1 | NP_001304884.1 | ||
| NM_001317956.1 | NP_001304885.1 | |||
| NM_001317957.1 | NP_001304886.1 | |||
| NM_004508.3 | NP_004499.2 | |||
| NR_134300.1 | ||||
| NR_134301.1 | ||||
| XM_011519457.1 | XP_011517759.1 | |||
| XM_017016191.1 | XP_016871680.1 | |||
| AK222875.1 | BAD96595.1 | |||
| AK303669.1 | ||||
| AK311110.1 | ||||
| BC057827.1 | AAH57827.1 | |||
| BC107893.1 | AAI07894.1 | |||
| BE891119.1 | ||||
| BI600349.1 | ||||
| BX537663.1 | 2 | 1395 | ||
| CD251816.1 | ||||
| CN481934.1 | ||||
| DA289206.1 | ||||
| DA792902.1 | ||||
| DA842794.1 | ||||
| DB479813.1 | ||||
| X17025.1 | CAA34890.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion |
|
| nsv5532 | Chr.10:1033559 - 1055760 on Build GRCh38 | Deletion |
|
| nsv1046220 | Chr.10:1018084 - 1143058 on Build GRCh38 | Gain |
|
| esv3891735 | Chr.10:978274 - 1064524 on Build GRCh38 | Gain |
|
| nsv549765 | Chr.10:688289 - 1083587 on Build GRCh38 | Gain |
|
| nsv951231 | Chr.10:1037361 - 1046360 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map