Assay Details
Target Gene Details
Entrez Gene ID: | 6585 |
Gene Name: | slit guidance ligand 1 |
Gene Aliases: |
MEGF4, SLIL1, SLIT-1, SLIT3 |
Location: |
Chr.10:96998038-97185926 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLIT1 | NM_003061.2 | NP_003052.2 | ||
| AB011537.3 | BAA32465.3 | |||
| AB017167.1 | BAA35184.1 | |||
| BC146761.1 | ||||
| BC146851.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052931 | Chr.10:96916435 - 97102544 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map