Assay Details
Target Gene Details
Entrez Gene ID: | 441549 |
Gene Name: | cerebral dopamine neurotrophic factor |
Gene Aliases: |
ARMETL1 |
Location: |
Chr.10:14819245-14838073 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDNF | NM_001029954.2 | NP_001025125.2 | ||
| XM_011519488.2 | XP_011517790.1 | |||
| BC037872.1 | AAH37872.1 | |||
| BC133042.1 | ||||
| BC133044.1 | ||||
| CN311321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471664 | Chr.10:14676176 - 14826378 on Build GRCh38 | Gain+Loss |
|
| nsv1043133 | Chr.10:14711571 - 14869583 on Build GRCh38 | Loss |
|
| nsv5899 | Chr.10:14803505 - 14837648 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79004966] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map