Assay Details
Target Gene Details
Entrez Gene ID: | 221037 |
Gene Name: | jumonji domain containing 1C |
Gene Aliases: |
TRIP-8, TRIP8 |
Location: |
Chr.10:63167221-63522075 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2677353 | Chr.10:63130283 - 63501631 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map