Assay Details
Target Gene Details
Entrez Gene ID: | 6832 |
Gene Name: | Suv3 like RNA helicase |
Gene Aliases: |
SUV3 |
Location: |
Chr.10:69180204-69209098 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUPV3L1 | NM_001301683.1 | NP_001288612.1 | ||
| NM_001323584.1 | NP_001310513.1 | |||
| NM_001323585.1 | NP_001310514.1 | |||
| NM_001323586.1 | NP_001310515.1 | |||
| NM_001323587.1 | NP_001310516.1 | |||
| NM_001323588.1 | NP_001310517.1 | |||
| NM_003171.4 | NP_003162.2 | |||
| NR_136626.1 | ||||
| NR_136627.1 | ||||
| AF042169.1 | AAB97370.1 | |||
| AK290416.1 | ||||
| AK299670.1 | ||||
| BC036112.1 | AAH36112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508589 | Chr.10:69166011 - 69276277 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map