Assay Details
Target Gene Details
Entrez Gene ID: | 6229 |
Gene Name: | ribosomal protein S24 |
Gene Aliases: |
DBA3, S24 |
Location: |
Chr.10:78033760-78056813 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS24 | NM_001142285.1 | 5 | 2215 | NP_001135757.1 |
More Information
Set Membership: |
|

Genomic Map