Assay Details
Target Gene Details
Entrez Gene ID: | 100506939 |
Gene Name: | PRKG1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:52296848-52314128 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKG1-AS1 | NR_038277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045692 | Chr.10:51541690 - 52327063 on Build GRCh38 | Gain |
|

Genomic Map