Assay Details
Target Gene Details
Entrez Gene ID: | 6441 |
Gene Name: | surfactant protein D |
Gene Aliases: |
COLEC7, PSP-D, SFTP4, SP-D |
Location: |
Chr.10:79937740-79982236 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFTPD | XM_011540087.1 | XP_011538389.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551635 | Chr.10:79925268 - 80212630 on Build GRCh38 | Loss |
|
| esv3623933 | Chr.10:79923186 - 80075168 on Build GRCh38 | Loss |
|
| nsv520754 | Chr.10:79955982 - 79980164 on Build GRCh38 | Loss |
|
| dgv941n100 | Chr.10:79884324 - 80099035 on Build GRCh38 | Gain |
|
| esv3623930 | Chr.10:79876627 - 79978510 on Build GRCh38 | Loss |
|
| nsv467391 | Chr.10:79881442 - 79980164 on Build GRCh38 | Gain |
|
| nsv831922 | Chr.10:79921025 - 80074699 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77565539] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map