Assay Details
Target Gene Details
Entrez Gene ID: | 10006 |
Gene Name: | abl interactor 1 |
Gene Aliases: |
ABI-1, ABLBP4, E3B1, NAP1BP, SSH3BP, SSH3BP1 |
Location: |
Chr.10:26746596-26861087 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831812 | Chr.10:26772577 - 26945338 on Build GRCh38 | Gain |
|
| esv3415799 | Chr.10:26640952 - 26944612 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map