Assay Details
Target Gene Details
Entrez Gene ID: | 5654 |
Gene Name: | HtrA serine peptidase 1 |
Gene Aliases: |
ARMD7, CADASIL2, CARASIL, HtrA, L56, ORF480, PRSS11 |
Location: |
Chr.10:122461525-122514908 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTRA1 | NM_002775.4 | NP_002766.1 | ||
| AF070555.1 | ||||
| AF097709.1 | AAC97211.1 | |||
| AK091944.1 | ||||
| AK092476.1 | ||||
| AK290089.1 | ||||
| BC011352.1 | ||||
| D87258.1 | BAA13322.1 | |||
| Y07921.1 | CAA69226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7580 | Chr.10:122446276 - 122490980 on Build GRCh38 | Deletion |
|
| nsv825594 | Chr.10:122434264 - 122505709 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map