Assay Details
Target Gene Details
Entrez Gene ID: | 23555 |
Gene Name: | tetraspanin 15 |
Gene Aliases: |
2700063A19Rik, NET-7, NET7, TM4SF15 |
Location: |
Chr.10:69451464-69549508 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN15 | NM_012339.3 | NP_036471.1 | ||
| XM_005269667.3 | XP_005269724.1 | |||
| XM_017016010.1 | XP_016871499.1 | |||
| AF120266.1 | AAD17295.1 | |||
| BC003157.1 | AAH03157.1 | |||
| BC004161.2 | AAH04161.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3623745 | Chr.10:69443738 - 69453601 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115158507] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map