Assay Details
Target Gene Details
Entrez Gene ID: | 55506 |
Gene Name: | H2A histone family member Y2 |
Gene Aliases: |
macroH2A2 |
Location: |
Chr.10:70052601-70112284 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2AFY2 | NM_018649.2 | NP_061119.1 | ||
| AF151534.1 | AAF72101.1 | |||
| AF336304.1 | AAK52471.1 | |||
| AK022475.1 | BAB14049.1 | |||
| AK022776.1 | BAB14239.1 | |||
| AK131057.1 | ||||
| BC016172.2 | AAH16172.1 | |||
| CX752382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
|
| nsv551479 | Chr.10:70007598 - 70179618 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74139258] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map