Assay Details
Target Gene Details
Entrez Gene ID: | 1787 |
Gene Name: | tRNA aspartic acid methyltransferase 1 |
Gene Aliases: |
DMNT2, DNMT2, MHSAIIP, PUMET, RNMT1 |
Location: |
Chr.10:17136785-17202071 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831801 | Chr.10:17085685 - 17265571 on Build GRCh38 | Gain |
|
| nsv1039697 | Chr.10:17006669 - 17549229 on Build GRCh38 | Loss |
|
| nsv1045115 | Chr.10:17037131 - 17167193 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map