Assay Details
Target Gene Details
Entrez Gene ID: | 219770 |
Gene Name: | gap junction protein delta 4 |
Gene Aliases: |
CX40.1 |
Location: |
Chr.10:35605410-35608935 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJD4 | NM_153368.2 | NP_699199.2 | ||
| AJ414564.1 | CAC93846.1 | |||
| AK074504.1 | BAC11028.1 | |||
| BC035898.1 | 1 | 1232 | AAH35898.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv144n67 | Chr.10:35600010 - 35681917 on Build GRCh38 | Gain |
|
| nsv466871 | Chr.10:35600599 - 35673959 on Build GRCh38 | Loss |
|
| esv3622902 | Chr.10:35606270 - 35610297 on Build GRCh38 | Loss |
|
| nsv825337 | Chr.10:35583021 - 35685326 on Build GRCh38 | Gain |
|
| esv2659329 | Chr.10:35606317 - 35610220 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map