Assay Details
Target Gene Details
Entrez Gene ID: | 221035 |
Gene Name: | receptor accessory protein 3 |
Gene Aliases: |
C10orf74, Yip2b |
Location: |
Chr.10:63521353-63625123 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REEP3 | NM_001001330.2 | NP_001001330.1 | ||
| XM_011539501.2 | XP_011537803.1 | |||
| XM_017015896.1 | XP_016871385.1 | |||
| AK289976.1 | ||||
| AY562241.1 | AAT70686.1 | |||
| BC010040.2 | AAH10040.1 | |||
| BC057832.1 | AAH57832.1 | |||
| BC068557.1 | AAH68557.1 | |||
| KJ534927.1 | ||||
| KJ535017.1 | ||||
| KJ535031.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551184 | Chr.10:63501424 - 63719655 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72837089] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map