Assay Details
Target Gene Details
Entrez Gene ID: | 220930 |
Gene Name: | ZEB1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:31316528-31319095 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZEB1-AS1 | NR_024284.1 | 2 | 1650 | |
| AK026657.1 | 1 | 1610 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831824 | Chr.10:31290791 - 31340754 on Build GRCh38 | Loss |
|

Genomic Map