Assay Details
Target Gene Details
Entrez Gene ID: | 132 |
Gene Name: | adenosine kinase |
Gene Aliases: |
AK |
Location: |
Chr.10:74151185-74709303 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADK | NM_001123.3 | NP_001114.2 | ||
| NM_001202449.1 | NP_001189378.1 | |||
| NM_001202450.1 | NP_001189379.1 | |||
| NM_006721.3 | NP_006712.2 | |||
| XM_017015698.1 | XP_016871187.1 | |||
| XM_017015699.1 | XP_016871188.1 | |||
| XM_017015700.1 | XP_016871189.1 | |||
| XM_017015701.1 | XP_016871190.1 | |||
| XM_017015702.1 | XP_016871191.1 | |||
| XM_017015703.1 | XP_016871192.1 | |||
| XM_017015704.1 | XP_016871193.1 | |||
| XM_017015705.1 | XP_016871194.1 | |||
| XM_017015706.1 | XP_016871195.1 | |||
| AK290633.1 | ||||
| AK301590.1 | ||||
| AK302706.1 | ||||
| BC003568.1 | AAH03568.1 | |||
| BT006788.1 | AAP35434.1 | |||
| U33936.1 | AAB01689.1 | |||
| U50196.1 | AAA97893.1 | |||
| U90338.1 | AAB50234.1 | |||
| U90339.1 | AAB50235.1 |
Target Gene Details
Entrez Gene ID: | 102723439 |
Gene Name: | uncharacterized LOC102723439 |
Gene Aliases: |
- |
Location: |
Chr.10:74506498-74529324 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102723439 | NR_120673.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1266n54 | Chr.10:74477740 - 74614146 on Build GRCh38 | Loss |
|
| nsv551528 | Chr.10:74332421 - 74600264 on Build GRCh38 | Loss |
|
| nsv1046267 | Chr.10:74472784 - 74535121 on Build GRCh38 | Gain |
|
| nsv515824 | Chr.10:74163688 - 74699911 on Build GRCh38 | Gain+Loss |
|
| dgv112n27 | Chr.10:74487347 - 74614146 on Build GRCh38 | Loss |
|
| dgv1265n54 | Chr.10:74386136 - 74600264 on Build GRCh38 | Loss |
|
| nsv7165 | Chr.10:74494436 - 74523578 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map