Assay Details
Target Gene Details
Entrez Gene ID: | 101929145 |
Gene Name: | PPP3CB antisense RNA 1 (head to head) |
Gene Aliases: |
USP54-AS1 |
Location: |
Chr.10:73495752-73507309 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP3CB-AS1 | NR_132103.1 | 5 | 2013 | |
| NR_132104.1 | 5 | 1540 |
Target Gene Details
Entrez Gene ID: | 159195 |
Gene Name: | ubiquitin specific peptidase 54 |
Gene Aliases: |
C10orf29, bA137L10.3, bA137L10.4 |
Location: |
Chr.10:73497538-73626844 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
More Information
Set Membership: |
|

Genomic Map