Assay Details
Target Gene Details
Entrez Gene ID: | 2674 |
Gene Name: | GDNF family receptor alpha 1 |
Gene Aliases: |
GDNFR, GDNFRA, GFR-ALPHA-1, RET1L, RETL1, TRNR1 |
Location: |
Chr.10:116056925-116273645 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFRA1 | NM_001145453.2 | NP_001138925.1 | ||
| NM_005264.5 | NP_005255.1 | |||
| NM_145793.3 | NP_665736.1 | |||
| XM_005269696.1 | XP_005269753.1 | |||
| XM_011539634.1 | XP_011537936.1 | |||
| AF038421.1 | AAC39692.1 | |||
| AF042080.1 | AAB97371.1 | |||
| AK292886.1 | ||||
| AK302914.1 | ||||
| BC014962.1 | AAH14962.1 | |||
| BU935138.1 | ||||
| U95847.1 | AAB71811.1 | |||
| U97144.1 | AAC51646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv975067 | Chr.10:116076369 - 116079842 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map