Assay Details
Target Gene Details
Entrez Gene ID: | 288 |
Gene Name: | ankyrin 3, node of Ranvier (ankyrin G) |
Gene Aliases: |
ANKYRIN-G, MRT37 |
Location: |
Chr.10:60026298-60733526 on Build GRCh38 |
Assay Gene Location: | Within Exon 52 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANK3 | NM_001149.3 | 21 | 5986 | NP_001140.2 |
| NM_001204403.1 | 44 | 8124 | NP_001191332.1 | |
| NM_001204404.1 | 44 | 8367 | NP_001191333.1 | |
| NM_001320874.1 | 43 | 8381 | NP_001307803.1 | |
| NM_020987.4 | 44 | 15911 | NP_066267.2 |
More Information
Set Membership: |
|

Genomic Map