Assay Details
Target Gene Details
Entrez Gene ID: | 1355 |
Gene Name: | COX15, cytochrome c oxidase assembly homolog |
Gene Aliases: |
CEMCOX2 |
Location: |
Chr.10:99694793-99732667 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX15 | NM_001320974.1 | NP_001307903.1 | ||
| NM_001320975.1 | 8 | 4929 | NP_001307904.1 | |
| NM_001320976.1 | 9 | 5001 | NP_001307905.1 | |
| NM_004376.6 | 9 | 3845 | NP_004367.2 | |
| NM_078470.5 | 9 | 5084 | NP_510870.1 | |
| XM_006717634.3 | XP_006717697.1 | |||
| XM_017015715.1 | XP_016871204.1 | |||
| AI760159.1 |
More Information
Set Membership: |
|

Genomic Map